Would you like to log in again?
Bacto™ Proteose Peptone No. 3, Gibco™ (a part of Thermo Fisher Scientific)
Supplier: Thermo Fisher Scientific
Bacto™
212230 212230 211693 212220 211692 212220 211693 211692
76628-782EA 1764.51 USD InStock
CA76628-928 76628-782 CA76628-782 76628-922 CA76628-922 76628-858 CA76628-858 76628-928
Bacto™ Proteose Peptone No. 3, Gibco™ (a part of Thermo Fisher Scientific)
Media for Microbiology Dehydrated Media for Microbiology
Bacto Proteose Peptone No. 3 is a member of the Bacto Proteose Peptones, which are enzymatic digests of protein.
  • Bacto Proteose Peptone is used in preparing microbiological culture media and in producing bacterial toxins
  • Bacto Proteose Peptone No. 2 is used in preparing microbiological culture media
  • Bacto Proteose Peptone No. 3 is used in preparing microbiological and mammalian cell culture media
  • Bacto Proteose Peptone No. 4 is a spray-dried version of Bacto Proteose Peptone
Bacto Proteose Peptone No. 4 offers the same beneficial nutrients as Bacto Proteose Peptone for growth promotion and toxin production in a wide range of fastidious microorganisms.
Certifications: LUO, MUO.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty