Would you like to log in again?
Cell mAb Medium, Quantum Yield, Gibco™ (a part of Thermo Fisher Scientific)
Supplier: Thermo Fisher Scientific
220511 220511
76628-924EA 60.11 USD InStock
76628-924 CA76628-924
Cell mAb Medium, Quantum Yield, Gibco™ (a part of Thermo Fisher Scientific)
Media for Microbiology Prepared Media for Microbiology
Cell MAb Medium, quantum yield, is a chemically defined (CD) basal cell culture medium. It contains L-glutamine and phenol red and does not contain Pluronic™ F68 (Kolliphor™ P188) or other surfactants.
  • Chemically defined (CD)
  • No pluronic acid and other surfactants
  • Contains L-glutamine and phenol red
Cell MAb Medium, quantum yield, should be supplemented in the same way as traditional basal medium (e.g., serum or serum-free supplements).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty