Would you like to log in again?
Gelysate™ Peptone, Gibco™ (a part of Thermo Fisher Scientific)
Supplier: Thermo Fisher Scientific
Gelysate™
211870 211870
76628-926EA 129.28 USD InStock
76628-926 CA76628-926
Gelysate™ Peptone, Gibco™ (a part of Thermo Fisher Scientific)
Media for Microbiology Dehydrated Media for Microbiology
Gelysate Peptone is a pancreatic digest of gelatin, derived from porcine animal origin (AO) sources. Gelatin hydrolysate is high in proline residues. Gelysate Peptone is deficient in carbohydrates and is characterized by low cystine, methionine, and tryptophan content.
  • Cultivation of heterotrophic microorganisms
Certifications: LUO
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty