Would you like to log in again?
Bacto™ Casitone, Gibco™ (a part of Thermo Fisher Scientific)
Supplier: Gibco™ (a part of Thermo Fisher Scientific)
Bacto™
225910 225930 225930 225910
76628-880EA 198.44 USD InStock
76628-880 CA76628-880 76628-932 CA76628-932
Bacto™ Casitone, Gibco™ (a part of Thermo Fisher Scientific)
Media for Microbiology Additives and Supplements for Microbiology
Bacto Casitone is an animal origin (AO) pancreatic enzymatic digest of casein similar to Difco Casein Digest.
  • Pancreatic digest of casein
  • More nutritious hydrolysate, especially for those organisms that prefer peptides to amino acids
  • Can be used as a component in microbiological media or in fermentation applications
While this supplement provides a nutritional profile based on casein, the specific enzymatic digest performed for this product results in a particularly high content of amino acids and peptides of varying sizes, making it a more nutritious hydrolysate. Can be used for the production of Escherichia coli based recombinant proteins.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty