Would you like to log in again?
Cell mAb Medium, Animal Component Free, Gibco™ (a part of Thermo Fisher Scientific)
Supplier: Thermo Fisher Scientific
220513 220513
76628-942EA 98.66 USD InStock
76628-942 CA76628-942
Cell mAb Medium, Animal Component Free, Gibco™ (a part of Thermo Fisher Scientific)
Media for Microbiology Prepared Media for Microbiology
Cell MAb Medium, animal component free, is a complete medium.
  • Contains L-glutamine
  • Supplemented with 0.3% Difco Select Soytone, and enzymatic digest of soy
  • No phenol red, pluronic acid or other surfactants
It does not contain attachment factors, which may need to be added if this medium is used with adherent cell lines.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty