Would you like to log in again?
Wheat Peptone 100, Ultra-Filtered (UF), Gibco™ (a part of Thermo Fisher Scientific)
Supplier: Thermo Fisher Scientific
670140 670139 670140 670139
76628-950EA 282.66 USD InStock
76628-950 CA76628-950 76628-868 CA76628-868
Wheat Peptone 100, Ultra-Filtered (UF), Gibco™ (a part of Thermo Fisher Scientific)
Media for Microbiology Dehydrated Media for Microbiology
Wheat 100, Ultra-Filtered (UF), is hydrolyzed wheat gluten protein that has been digested for enhanced peptide content, resulting in a low concentration of free amino acids and carbohydrates. It is an ultra-filtered supplement and is ideal for mammalian cell culture applications.
Wheat 100, UF, is also part of the Peptone Preview Pak which contains:
Cotton 100, UF - rich in nucleoside content and digested to enhance peptide content. It is processed to provide higher concentrations of vitamins, which can help maintain performance in mammalian and microbial processes.
Wheat 100, UF - ultra-filtered supplement and good source of glutamine, which makes it ideal for mammalian cell culture applications. It is digested for enhanced peptide content resulting in a low concentraion of free amino acids and carbohydrates.
Soy 100 - highly digested soy protein peptone that is high in nucleosides and free amino acid content. It has high concentrations of vitamins and carbohydrates and is recommended for a wide variety of processes.
Pea 100, UF - ultra-filtered peptone digested to enhance nucleoside and free amino acid content. The high total amino acid content and low carbohydrate content are similar to many high-performance animal origin peptones.
Yeast 100 - designed to enhance the content of vitamins and other key nutritional components found in yeast. It is intended for nutritional supplementation for mammalian, microbial, and insect cell culture.
Wheat 100, UF, contains glutamine that is associated with peptides, making it a stable source. This helps minimize the ammonia generation compared to the addition of free glutamine in cell culture media. Wheat 100, UF, can also be used to enhance growth and production in microbial applications.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty