Would you like to log in again?
Bacto™ Proteose Peptone No. 4, Gibco™ (a part of Thermo Fisher Scientific)
Supplier: Thermo Fisher Scientific
Bacto™
211715 211715
76628-964EA 4360.23 USD InStock
76628-964 CA76628-964
Bacto™ Proteose Peptone No. 4, Gibco™ (a part of Thermo Fisher Scientific)
Media for Microbiology Dehydrated Media for Microbiology
Bacto Proteose Peptone No. 4 is a member of the Bacto Proteose Peptones, which are enzymatic digests of protein.
  • Bacto Proteose Peptone is used in preparing microbiological culture media and in producing bacterial toxins
  • Bacto Proteose Peptone No. 2 is used in preparing microbiological culture media
  • Bacto Proteose Peptone No. 3 is used in preparing microbiological and mammalian cell culture media
  • Bacto Proteose Peptone No. 4 is a spray-dried version of Bacto Proteose Peptone
It offers the same beneficial nutrients as Bacto Proteose Peptone for growth promotion and toxin production in a wide range of fastidious microorganisms.
Studies of peptic digests of animal tissue prepared under varying digestion parameters demonstrated that no single peptone is the most suitable nitrogen source for every microbiological application. These findings led to the development of Bacto Proteose Peptone, Bacto Proteose Peptone No. 2, and Bacto Proteose Peptone No. 3. Bacto Proteose Peptone No. 4 is a spray-dried version of Bacto Proteose Peptone.
We also offer alternatives to Bacto Proteose Peptones for scale-up to production applications, including BiTek Proteose Peptone.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty