Would you like to log in again?
RayDyLyo® Sterile Snap Caps, DWK Life Sciences
Supplier: DWK Life Sciences
RayDyLyo®
255853000 255843001 255845002 255853002 255855003 255845003 255855001 255845001 255854004 255843002 255853001 255853003 255845000 255843000 255843003 255854001
76632-788CS 6495.94 USD InStock
76632-788 76632-810 76632-802 76632-824 76632-826 76632-828 76632-818 76632-808 76632-790 76632-780 76632-782 76632-794 76632-784 76632-830 76632-786 76632-820
RayDyLyo® Sterile Snap Caps, DWK Life Sciences
Closures Chromatography Vial Closures
RayDyLyo® plastic push-fit caps.
  • Flip off snap caps are pre-assembled with an ISO 8362-2 compliant elastomer stopper
  • Container closure integrity (CCI) validated for use with ISO 8362-1 glass and COP vials
  • Delivered sterile (RTU) with a 2-year shelf life from time of manufacturer
  • Clean room manufactured with 100% in-process quality controls
  • Suitable for aseptic small batch, clinical trial, and robotic fill and finish operations
  • Snap caps do not require special crimping tools and affix with single vertical pressure
  • Provides tamper-evident closing and eliminates crimping defects
  • Compatible with gamma and steam terminal sterilization
  • Available in nested, standard bag, and RTP-type bag configurations
RayDyLyo® plastic push-fit caps from DWK Life Sciences, designed for closing vials of injectable drugs. These flip-off snap caps are pre-assembled with an ISO 8362-2 compliant elastomer stopper to simplify the production process.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty