Would you like to log in again?
Primo™ Laser Histology Cassette Printer, Azer Scientific
Supplier: Azer Scientific
ES39915
76632-836EA 42860.4 USD InStock
76632-836
Primo™ Laser Histology Cassette Printer, Azer Scientific
Printers Cassette and Slide Printers
Azer Primo™ Cassette Printer allows for quick, permanent and efficient printing on histology tissue and biopsy cassettes. UV laser technology eliminates the need of expensive printer consumables and maintenance.
  • Laser tissue cassettes printer (ribbon or cartridge free printing)
  • Six cassette magazines hold up to 480 cassettes total, load six different color cassettes for efficient workflow
  • Precision construction for jam-free cassette printing
  • 1D and 2D barcodes, alphanumeric, graphic and logo printing
  • Low work noise ≤65 dB, meet the hospital pathology and labs environment requirements
  • Easily Integrate to LIS/HIS system
  • High printing speed completes a cassette every 3 seconds
Our unique design eliminates the need for expensive ribbons, cartridges or print heads to optimize your workflow. Our printer will effortlessly connect to your laboratory and hospital information systems and is perfectly compatible with our hinged tissue cassettes with or without lids.
Ordering information: Two year warranty.
Delivery information: Includes two pre-installed templates. Contact prior to shipping for data fields required.
Caution: Do not attempt to open during operation.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty