Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

FLUIDSHIELD* Surgical N95 Respirator Masks, Halyard

Supplier: Halyard
Fluidshield
46727 46727
76632-880CS 835.38 USD InStock
76632-880 CA76632-880
FLUIDSHIELD* Surgical N95 Respirator Masks, Halyard
Respirators Disposable Respirators Particulate Respirators
FLUIDSHIELD* N95 filter respirators are NIOSH approved and are designed to provide an effective facial fit as required for proper respiratory protection.

  • NIOSH approved to filter out at least 95% of very small (<0.3 µm) particles, including bacteria and viruses
  • Bonded straps are not stapled to the mask
  • Malleable, adjustable nose wire for customized fit
  • Duckbill breathing chamber is more than twice as large as the leading competitive surgical N95
  • SO SOFT* inner lining for comfort
  • Compact, easy storage; folds flat without crushing
  • Meets and exceeds NIOSH breathability standards

Masks feature 5-layer construction with a soft inner lining. 100% natural rubber, latex-free, and available with safety seal.

100% of FLUIDSHIELD* N95 Respirators have been cleared by the FDA as a medical device, unlike many N95s or KN95s that have received a temporary Emergency Use Authorization.

Certifications: NIOSH approved. FDA cleared Class 2 medical devices, Level 3.

Packaging: 35 masks per box, 6 boxes per case, 210 masks per case.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty