Would you like to log in again?
HyClone™ VaccineXpress Cell Culture Media, Cytiva
Supplier: HyClone products (Cytiva)
HyClone™ VaccineXpress
SH31126.01 SH31126.01
76633-018EA 134.28 USD InStock
CA76633-018 76633-018
HyClone™ VaccineXpress Cell Culture Media, Cytiva
Cell Culture Media
HyClone VaccineXpress cell culture medium is designed and developed for high‑density growth and maintenance of kidney‑derived cell lines (e.g., Vero cells) for viral vaccine development. VaccineXpress medium is well suited for use in multi-well plates and large‑scale manufacturing using microcarriers in WAVE Bioreactor or Xcellerex bioreactors.
  • Serum-free animal-derived component-free formulation
  • Supports direct or sequential adaptation, cell growth and virus propagation
  • Designed to support high peak cell density, viral infectivity, and productivity
  • For use with multiple viral applications (i.e., influenza, rotavirus, RSV, Dengue, Zika)
  • Designed for small- to large-scale cell culture applications with use of microcarriers
  • Available in liquid and powder formulations
HyClone VaccineXpress medium has been qualified to grow attachment‑dependent Vero cells with microcarriers for producing viral vaccines. The lack of both serum and animal‑derived components in the medium reduces variability and enhances scalability and ease of purifying recombinant proteins and viruses in bioprocess applications. VaccineXpress is formulated without L‑glutamine for extended shelf life.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty