Would you like to log in again?
HyClone™ ActiSM™ Cell Culture Media, Cytiva
Supplier: Cytiva
HyClone™ ActiSM™
SH31040.02 SH31040.02
76633-020EA 122.17 USD InStock
CA76633-020 76633-020
HyClone™ ActiSM™ Cell Culture Media, Cytiva
Cell Culture Media
ActiSM™ cell culture media is a chemically defined animal-derived component-free (ADCF) cultured media optimized for high-yield protein production for bioprocessing. This HyClone™ medium is designed for use with ActiPro™ cell culture media.
  • Chemically defined ADCF formulation optimized for high-yield protein production
  • Does not contain hypoxanthine, thymidine, and glycine (HAT supplement)
  • Recommended for first step when adapting cells cultured in other lean cell culture media
ActiSM™ medium is intended for use in combination with ActiPro™ medium and Cell Boost™ 7a and Cell Boost™ 7b supplements. This HyClone™ medium is part of the ActiPro™ system of media and supplements designed with adaptation and production in mind. ActiSM™ and the ActiPro™ system are suitable for general biomanufacturing with CHO cell lines such as CHO-GS, CHO-K1, CHO-DG44, and CHO-S. Once your process has been adapted for use with the ActiSM™ medium, we recommend utilizing ActiPro™ medium for production.
ActiSM™ cell culture medium is available in both powder and liquid formats. ActiSM™ medium does not contain any growth factors (such as insulin), peptides, hydrolysates, phenol red, or 2-mercaptoethanol, ensuring CHO batch-to-batch consistency.
Cell culture media formulations and delivery systems can be customized to your specific process requirements or optimized to maximize process yields. Our cell culture media and development service is available to help you analyze, develop, and optimize your cell culture strategies.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty