Would you like to log in again?
PermaRed/HRP, Diagnostic BioSystems
Supplier: Diagnostic BioSystems
K075 K075
76633-066EA 150.48 USD InStock
CA76633-066 76633-066
PermaRed/HRP, Diagnostic BioSystems
Assays Immunological Assays
PermaRed/HRP is a substrate-chromogen system designed to be used for either IHC or ISH when utilizing horseradish peroxidase. PermaRed/HRP produces a distinct brick red stain that can easily be distinguished from other stains.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty