Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Anti-CD45RO T cell Mouse Monoclonal Antibody [clone: UCHL-1]
Supplier: Diagnostic BioSystems
MOB043-05-RUO MOB043-01 MOB043 MOB043-RUO MOB043 MOB043-01-RUO
76633-144EA 451.44 USD InStock
CA76633-144 CA76633-142 76633-144 CA76633-140 76633-142 76633-140
Anti-CD45RO T cell Mouse Monoclonal Antibody [clone: UCHL-1]
Antibodies
This antibody reacts with a 180 kDa glycoprotein of the CD45 family, found on most thymocytes and activated T cells but only on a portion of the resting T cells. It reacts with most thymocytes, a subpopulation of resting cells within both the CD4 and CD8 subsets and mature activated T cells.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty