Would you like to log in again?
Anti-MGMT Mouse Monoclonal Antibody [clone: MT3.1]
Supplier: Diagnostic BioSystems
MOB423-01-RUO MOB423-RUO MOB423-05 MOB423-RUO MOB423-05-RUO MOB423-01
76633-862EA 136.16 USD InStock
76633-862 CA76633-866 76633-866 CA76633-864 76633-864 CA76633-862
Anti-MGMT Mouse Monoclonal Antibody [clone: MT3.1]
Antibodies
O(6)-methylguanine-DNA-methyltransferase (MGMT) is a 22 kDa human DNA repair protein that removes O(6)-alkylguanine DNA adducts. MGMT acts as a suicide acceptor protein repairing the DNA by accepting alkyl groups and consequently inactivating itself. This antibody stains all mantle zone lymphocytes and 50% of germinal center lymphocytes. Basaloid epithelial cells of tonsil squamous mucosa are also stained positive with this antibody.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty