Would you like to log in again?
Anti-Cytokeratin 5 and 14 Mouse Monoclonal Antibody [clone: XM26 + LL002]
Supplier: Diagnostic BioSystems
MOB433-01-RUO MOB433-01 MOB433-05-RUO MOB433 MOB433-RUO MOB433-05
76633-902EA 693.88 USD InStock
CA76633-902 CA76633-900 76633-902 CA76633-898 76633-900 76633-898
Anti-Cytokeratin 5 and 14 Mouse Monoclonal Antibody [clone: XM26 + LL002]
Antibodies
This antibody is specific to 50 kDa cytokeratin protein designated cytokeratin 14. Cytokeratin 14 belongs to the type A (acidic) subfamily of low molecular weight cytokeratins and exists in combination with cytokeratin 15. This antibody can be used to distinguish stratified epithelial cells from simple epithelial cells.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty