Would you like to log in again?
Anti-BCL10 Mouse Monoclonal Antibody [clone: 151]
Supplier: Diagnostic BioSystems
MOB442-05 MOB442-01 MOB442 MOB442-01-RUO MOB442-RUO MOB442-05-RUO
76633-926EA 530.27 USD InStock
CA76633-926 CA76633-924 76633-926 CA76633-922 76633-924 76633-922
Anti-BCL10 Mouse Monoclonal Antibody [clone: 151]
Antibodies
Monoclonal Anti-BCL-10 reacts specifically with human bcl-10. The epitope recognized by the antibody resides within amino acids 122-168 of human bcl-10 molecule. BCL-10, also designated as CIPER, mE10, cE10, CARMEN, and CLAP, is an N-terminal CARD (caspase recruitment domain) containing protein. It is a cellular homologue of the equine herpesvirus-2 protein E-10 (VCLAP). BCL-10 was implicated in the regulation of apoptosis by interacting with caspase 9, enhancing procaspase 9 processing, and triggering its activation when overexpressed in the cell. Bcl-10 cellular overexpression induces JNK, P38, and NF-kB activation. Deregulation of BCL-10 expression was also demonstrated to be involved in cellular oncogenesis.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty