Would you like to log in again?
Anti-Cytokeratin 13 Mouse Monoclonal Antibody [clone: KS-1A3]
Supplier: Diagnostic BioSystems
MOB443 MOB443-RUO MOB443-01 MOB443-01-RUO MOB443-05 MOB443-05-RUO
76633-928EA 105.1 USD InStock
CA76633-928 76633-928 CA76633-932 76633-932 CA76633-930 76633-930
Anti-Cytokeratin 13 Mouse Monoclonal Antibody [clone: KS-1A3]
Antibodies
This antibody is specific to 54 kDa protein. Cytokeratin 13 belongs to the type A (acidic) subfamily of low molecular weight cytokeratins and exists in combination with cytokeratin 4. Cytokeratin 13 is expressed in squamous, non-keratinized epithelium, transitional epithelium and myoepithelium. This antibody is useful for the identification of carcinomas of trachea, sweat glands, bladder, ectocervix, tongue, esophagus, anal canal and the basal layer of keratinized epidermis.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty