Would you like to log in again?
Anti-Napsin A Mouse Monoclonal Antibody [clone: KCG1.1]
Supplier: Diagnostic BioSystems
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
MOB463-05-RUO MOB463-01 MOB463-01-RUO MOB463-RUO MOB463-05 MOB463
76633-956EA 653.28 USD InStock
CA76633-956 76633-956 CA76633-954 76633-954 CA76633-952 76633-952
Anti-Napsin A Mouse Monoclonal Antibody [clone: KCG1.1]
Antibodies
Napsin is found in two isoforms, A and B, with highly homologous gene sequences. Napsin A has a molecular weight of 35.0 kDa and is also known as TA02. Napsin A is an aspartic proteinase which is expressed in the lung and involved in processing surfactant protein B (SP-B). It is also expressed in the kidney. This antibody may be a useful tool as a tumor marker for primary lung adenocarcinoma. Napsin expression correlates with the differentiation grade of lung adenocarcinoma.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty