Would you like to log in again?
Anti-Renal Cell Carcinoma Mouse Monoclonal Antibody [clone: PN-15]
Supplier: Diagnostic BioSystems
MOB465 MOB465-05-RUO MOB465-RUO MOB465-01-RUO MOB465-05 MOB465-01
76633-968EA 541.01 USD InStock
CA76633-968 CA76633-966 76633-968 CA76633-964 76633-966 76633-964
Anti-Renal Cell Carcinoma Mouse Monoclonal Antibody [clone: PN-15]
Antibodies
This antibody recognizes a 200 kDa glycoprotein (GP200) localized in the brush border of the proximal renal tebules and the luminal surface of Bowman’s capsule. GP200 was expressed by 93% of primary and 84% of metastatic renal carcinomas. GP200 glycoprotein is also localized in the breast lobules and ducts, parathyroid glands and epididymis.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty