Would you like to log in again?
Anti-DOG1.1 Mouse Monoclonal Antibody [clone: DOG1.1]
Supplier: Diagnostic BioSystems
MOB466-RUO MOB466-05-RUO MOB466-01 MOB466-01-RUO MOB466 MOB466-05
76633-972EA 327.24 USD InStock
76633-972 CA76633-970 76633-970 CA76633-974 76633-974 CA76633-972
Anti-DOG1.1 Mouse Monoclonal Antibody [clone: DOG1.1]
Antibodies
DOG1 is expressed ubiquitously in gastrointestinal stromal tumors irrespective of c-kit or PDGFR alpha mutation status. Reactivity for DOG1 may aid in the diagnosis of gastrointestinal stromal tumors, including PDGFRA mutants that fail to express c-kit antigen, and lead to appropriate treatment with imatinib mesylate, an inhibitor of the c-kit tyrosine kinase.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty