Would you like to log in again?
Anti-Cytokeratin (CAM5.2) Mouse Monoclonal Antibody [clone: CAM5.2]
Supplier: Diagnostic BioSystems
MOB469-05-RUO MOB469-01 MOB469-05 MOB469-RUO MOB469 MOB469-01-RUO
76633-984EA 509.96 USD InStock
CA76633-982 76633-984 76633-982 CA76633-986 CA76633-984 76633-986
Anti-Cytokeratin (CAM5.2) Mouse Monoclonal Antibody [clone: CAM5.2]
Antibodies
Anti-Cytokeratin (CAM 5.2) reagent has a primary reactivity with human keratin proteins that correspond to Moll’s peptides #7 and #8, Mr 48 and 52 kilodaltons (kd), respectively. Cytokeratin 7 and 8 are present on secretory epithelia of normal human tissue but not onstratified squamous epithelium. Anti-Cytokeratin (CAM 5.2) stains most epithelial-derived tissue, including liver, renal tubular epithelium, and hepatocellular and renal cell carcinomas. Anti-Cytokeratin (CAM 5.2) might not react with some squamous cell carcinomas.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty