Would you like to log in again?
Anti-C4D Mouse Monoclonal Antibody [clone: C4D204]
Supplier: Diagnostic BioSystems
MOB471-RUO MOB471 MOB471-01-RUO MOB471-05-RUO MOB471-01 MOB471-05
76633-992EA 668.8 USD InStock
CA76633-992 76633-992 CA76633-990 76633-990 CA76633-988 76633-988
Anti-C4D Mouse Monoclonal Antibody [clone: C4D204]
Antibodies
Complement 4d (C4D) is the most clinically used marker for humoral rejection. It is a degradation product of the activated complement factor C4B. Complement 4D is typically initiated by binding of antibodies to specific target molecules. Following activation and degradation of the C4 molecule, thio-ester groups are exposed, which allow transient, covalent binding of the degradation product Complement 4d to endothelial cell surfaces and extracellular matrix components of vascular basement membranes near the sites of C4 activation. Complement 4D is also found in intracytoplasmic vacuoles of endothelial cells. Covalent binding renders C4D a stable molecule that can easily be detected by immunohistochemistry.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty