Would you like to log in again?
Anti-Tau (Neurofibrillary Tangles Marker) Mouse Monoclonal Antibody [clone: Tau46]
Supplier: Diagnostic BioSystems
MOB472-01-RUO MOB472-01 MOB472-05 MOB472-05-RUO MOB472 MOB472-RUO
76633-994EA 117.04 USD InStock
76633-994 CA76633-998 76633-998 CA76633-996 76633-996 CA76633-994
Anti-Tau (Neurofibrillary Tangles Marker) Mouse Monoclonal Antibody [clone: Tau46]
Antibodies
This antibody is specific to 45-60 kDa proteins identified as tau proteins. This antibody does not cross react with tubulin or other microtubule associated proteins. This antibody recognizes a phosphorylation-independent epitope in amino acids 404 to 441 (human). Tau antibody stains human neurofibrillary tangles, neutrophil threads and neurotic plaques associated with Alzheimer’s disease.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty