Would you like to log in again?
Anti-CD3 Mouse Monoclonal Antibody [clone: LN10]
Supplier: Diagnostic BioSystems
MOB474-05 MOB474-05-RUO MOB474 MOB474-01 MOB474-01-RUO MOB474-RUO
76634-002EA 549.37 USD InStock
CA76634-000 76634-002 CA76634-002 76634-004 76634-000 CA76634-004
Anti-CD3 Mouse Monoclonal Antibody [clone: LN10]
Antibodies
The clone F7.2.38 is specific for the non-glycosylated epsilon chain of the human CD3 molecule. Clone F7.2.38 recognizes T cells in the thymus, bone marrow, peripheral lymphoid tissue and blood and is a pan T cell marker. The CD3 molecule consists of five different polypeptide chains with molecular weights ranging from 16 to 28 kDa. The CD3 antigen is first detected in early thymocytes, and its appearance probably represents one of the earliest signs of commitment to the T cell lineage.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty