Would you like to log in again?
Anti-TdT Mouse Monoclonal Antibody [clone: SEN 28]
Supplier: Diagnostic BioSystems
MOB545-05 MOB545-05-RUO MOB545-01-RUO MOB545-01 MOB545 MOB545-RUO
76634-046EA 994.84 USD InStock
CA76634-044 76634-046 CA76634-046 76634-042 CA76634-042 76634-044
Anti-TdT Mouse Monoclonal Antibody [clone: SEN 28]
Antibodies
This antibody stains a 60 kDa peptide in human lymphoblasts. TdT is present in the cells during immunoglobulin gene rearrangement and plays an important role in lymphocyte maturation. TdT marks both T and B immature lymphocytes normally found in thymus and bone marrow.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty