Would you like to log in again?
Anti-MOC-31 Mouse Monoclonal Antibody [clone: MOC-31]
Supplier: Diagnostic BioSystems
MOB546-01-RUO MOB546-01 MOB546-RUO MOB546-05-RUO MOB546 MOB546-05
76634-048EA 115.85 USD InStock
76634-048 76634-052 CA76634-050 CA76634-052 76634-050 CA76634-048
Anti-MOC-31 Mouse Monoclonal Antibody [clone: MOC-31]
Antibodies
MOC-31 antibody recognizes an epithelial-associated, glycoprotein located on the cell membrane surface and in the cytoplasm of virtually all epithelial cells. It is not present in most squamous epithelia, hepatocytes, renal proximal tubular cells, gastric parietal cells and myoepithelial cells. MOC-31 may be used in a panel of antibodies as a negative marker for mesothelioma, or lung adenocarcinoma. Studies have shown that MOC31 is useful in differentiating tumors of unknown origin in liver cancers and distinguishing Cholangiocarcinoma from Hepatocellular Carcinomas.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty