Would you like to log in again?
Anti-CA 15-3 Mouse Monoclonal Antibody [clone: DBMM1]
Supplier: Diagnostic BioSystems
MOB549-RUO MOB549-01-RUO MOB549-01 MOB549-05 MOB549 MOB549-05-RUO
76634-064EA 771.52 USD InStock
CA76634-062 76634-064 CA76634-064 76634-060 CA76634-060 76634-062
Anti-CA 15-3 Mouse Monoclonal Antibody [clone: DBMM1]
Antibodies
This antibody is specific to epithelial membrane antigen (EMA), CA 15-3, or polymorphic epithelial mucin. This antibody stains an underglycosylated MUC1 often present on carcinoma cells.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty