Would you like to log in again?
Anti-E-Cadherin Mouse Monoclonal Antibody [clone: SPM471]
Supplier: Diagnostic BioSystems
MOB550-01-RUO MOB550-05-RUO MOB550-RUO MOB550-05 MOB550 MOB550-01
76634-068EA 474.13 USD InStock
CA76634-066 76634-068 CA76634-068 76634-066 76634-070 CA76634-070
Anti-E-Cadherin Mouse Monoclonal Antibody [clone: SPM471]
Antibodies
E-cadherin (uvomorulin, cell-CAM120/80) is a calcium dependent cell adhesion molecule expressed predominately in epithelial tissues. It plays an important role in the growth and development of cells via the mechanisms of control of tissue architecture and the maintenance of tissue integrity. Numerous studies have demonstrated that reduction and/or loss of E-cadherin expression in carcinomas correlates positively with the potential of these tumors for invasion and metastasis.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty