Would you like to log in again?
Anti-Calbindin Mouse Monoclonal Antibody [clone: CB-955]
Supplier: Diagnostic BioSystems
MOB554-01-RUO MOB554-05 MOB554 MOB554-RUO MOB554-01 MOB554-05-RUO
76634-078EA 105.1 USD InStock
76634-078 CA76634-078 CA76634-080 76634-082 CA76634-082 76634-080
Anti-Calbindin Mouse Monoclonal Antibody [clone: CB-955]
Antibodies
This antibody is specific to a protein of 28 kDa. Calbindin-D-28K is also known as vitamin D-dependent calcium-binding protein or cholecalcin. Calbindin D-28K has a broad tissue distribution; however, it exhibits a cell type-specific expression pattern. This antibody reacts specifically with calbindin-D-28K in brain and kidney tissues.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty