Would you like to log in again?
Anti-NKX3-1 Mouse Monoclonal Antibody [clone: 361]
Supplier: Diagnostic BioSystems
MOB569-05-RUO MOB569-01-RUO MOB569-05 MOB569-01 MOB569-RUO MOB569
76634-152EA 471.75 USD InStock
CA76634-154 CA76634-150 76634-152 CA76634-152 76634-154 76634-150
Anti-NKX3-1 Mouse Monoclonal Antibody [clone: 361]
Antibodies
The homeodomain-containing transcription factor NKX3-1 is a putative prostate tumor suppressor that is expressed in a largely prostate-specific and androgen-regulated manner. Loss of NKX3-1 protein expression is a common finding in human prostate carcinomas and prostatic intraepithelial neoplasia.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty