Would you like to log in again?
Anti-PD-L1 Mouse Monoclonal Antibody [clone: 405-9A11]
Supplier: Diagnostic BioSystems
MOB572-05-RUO MOB572-01 MOB572-RUO MOB572-05 MOB572-01-RUO MOB572-RUO
76634-166EA 655 USD InStock
76634-166 CA76634-164 CA76634-166 76634-162 76634-164 CA76634-162
Anti-PD-L1 Mouse Monoclonal Antibody [clone: 405-9A11]
Antibodies
The programmed death receptor 1 (PD-1) protein is a cell-surface receptor on certain lymphocytes that, with its ligand programmed death ligand 1 (PD-L1), helps to down-regulate immune responses. Many cancer types express PD-L1 and evade immune recognition via the PD-1/PD-L1 interaction. Precision therapies targeting the PD-1/PD-L1 pathway have the potential to improve response and thereby offer a novel treatment avenue to some patients with cancer.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty