Would you like to log in again?
Anti-Mismatch Repair Protein 2, MSH2 Mouse Monoclonal Antibody [clone: DBM15.82]
Supplier: Diagnostic BioSystems
MOB585-05-RUO MOB585-01-RUO MOB585-01-RUO MOB585-05 MOB585-RUO MOB585
76634-196EA 796.6 USD InStock
CA76634-194 76634-196 CA76634-196 76634-192 CA76634-192 76634-194
Anti-Mismatch Repair Protein 2, MSH2 Mouse Monoclonal Antibody [clone: DBM15.82]
Antibodies
Mutations in DNA mismatch repair genes are associated with hereditary nonpolyposis colorectal cancer (HNPCC). Initially, inherited mutations in the MSH2 and MLH1 homologs of the bacterial DNA mismatch repair genes MutS and MutL were found at high frequency in HNPCC and were shown to be associated with microsatellite instability. The demonstration that 10 to 45% of pancreatic, gastric, breast, ovarian and small cell lung cancers also display microsatellite instability has been interpreted to suggest that DNA mismatch repair is not restricted to HNPCC tumors but is a common feature in tumor initiation or progression.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty