Would you like to log in again?
Anti-BCL6 Mouse Monoclonal Antibody [clone: PG-B6]
Supplier: Diagnostic BioSystems
MOB587-01-RUO MOB587-RUO MOB587-01 MOB587-05-RUO MOB587-05-RUO MOB587-RUO
76634-208EA 663 USD InStock
CA76634-206 76634-208 CA76634-208 76634-204 CA76634-204 76634-206
Anti-BCL6 Mouse Monoclonal Antibody [clone: PG-B6]
Antibodies
The BCL6 gene encodes a 706 amino acid nuclear protein of the Kruppeltype zinc finger protein. It is rearranged in about 30% of diffuse large B-cell lymphomas, and is expressed predominantly in normal germinal centre B cells and related lymphomas. The antibody is a useful aid for classification of follicular lymphomas, diffuse large B-cell lymphomas, Burkitt's lymphomas, and nodular, lymphocyte-predominance Hodgkin's lymphoma. The BCL6 antibody, together with BCL2 antibody, is also a useful aid in classification of mantle cell lymphomas, and nodular, lymphocytepredominance Hodgkin's lymphoma. BCL6 protein is not expressed in BCLL, hairy cell leukemia, mantle cell and marginal-zone derived lymphomas
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty