Would you like to log in again?
Anti-Villin Mouse Monoclonal Antibody [clone: 1D2C3]
Supplier: Diagnostic BioSystems
MOB590-RUO MOB590-05-RUO MOB590-01-RUO MOB590-01-RUO MOB590-05-RUO MOB590
76634-222EA 133 USD InStock
76634-222 CA76634-222 76634-224 CA76634-224 76634-226 CA76634-226
Anti-Villin Mouse Monoclonal Antibody [clone: 1D2C3]
Antibodies
Villin is One of the gelsolin family of calcium regulated actin binding proteins. Normally expressed in the brush border of epithelial cells lining the gastrointestinal tract, hepatobiliary tract and renal proximal convoluted tubules. In colorectal carcinoma, villin is highly expressed. In practice, villin can be included in the panel used for metastatic carcinoma to detect colorectal origin.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty