Would you like to log in again?
Anti-Calretinin (H-5) Mouse Monoclonal Antibody [clone: H-5]
Supplier: Diagnostic BioSystems
MOB593-05 MOB593-01 MOB593-01-RUO MOB593-RUO MOB593 MOB593-05-RUO
76634-234EA 164.81 USD InStock
76634-234 CA76634-238 76634-236 CA76634-234 76634-238 CA76634-236
Anti-Calretinin (H-5) Mouse Monoclonal Antibody [clone: H-5]
Antibodies
Calbindin D28K and Calretinin (also designated CR or 29 kDa Calbindin) are two closely related intracellular calcium-binding proteins belonging to the Troponin-C superfamily. Initially isolated from chick retina. Calretinin is an intracellular calcium-binding protein belonging to the troponin C superfamily characterized by a structural motif described as the EF-hand domain. The immunohistochemical detection of calretinin in developing cerebellum is restricted to the later stages indicated by weak staining from week 21 of gestation in Purkinje and basket cells and in neurons of the dentate nucleus. The intensity of staining increases as the cerebellum matures. In tumors, calretinin has been detected in mesotheliomas and some pulmonary adenocarcinomas.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty