Would you like to log in again?
Anti-Uroplakin III (AU1) Mouse Monoclonal Antibody [clone: AU1]
Supplier: Diagnostic BioSystems
MOB594-01 MOB594-01-RUO MOB594-RUO MOB594-RUO MOB594-05 MOB594-05-RUO
76634-244EA 514 USD InStock
CA76634-242 76634-244 CA76634-244 76634-240 CA76634-240 76634-242
Anti-Uroplakin III (AU1) Mouse Monoclonal Antibody [clone: AU1]
Antibodies
Uroplakins (UPs) are a family of transmembrane proteins (UPs Ia, Ib, II and III) that are specific differentiation products of urothelial cells. In nonneoplastic mammalian urothelium, UPs are expressed in the luminal surface plasmalemma of superficial (umbrella) cells, forming complexes of 16nm crystalline particles. UPIII is specific for tumors of urothelial origin and, when used in combination with other markers, can aid in the diagnosis of primary and metastatic tumors.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty