Would you like to log in again?
Anti-A20 (A-12) Mouse Monoclonal Antibody [clone: A-12]
Supplier: Diagnostic BioSystems
MOB598-01 MOB598-01-RUO MOB598 MOB598-05-RUO MOB598-05 MOB598-RUO
76634-246EA 189.89 USD InStock
76634-246 76634-250 CA76634-250 CA76634-246 76634-248 CA76634-248
Anti-A20 (A-12) Mouse Monoclonal Antibody [clone: A-12]
Antibodies
A20 Antibody (A-12) is a high quality monoclonal A20 antibody (also designated TNFAIP3 antibody, or TNF a induced protein 3 antibody). A20 is induced by tumor necrosis factor (TNF) and interleukin 1 (IL-1) and acts as a negative regulator of nuclear factor kB (NFkB) gene expression. By inhibiting NFkB activation, A20 plays a critical role in terminating NFkB responses to various stimuli. Although the C-terminal region of A20 contains seven zinc finger domains, only four of these domains are required for in vitro inhibition of TNF-induced NFkB activation. A20 also interacts with several other proteins, such as TRAF2, TRAF6 and IkB kinase (IKK) g protein, and can thereby inhibit cell death. TXBP151, a novel A20-binding protein, may mediate the anti-apoptotic activity of A20. Involved in the negative feedback regulation of signal transduction, A20 and A20-binding proteins may be useful as novel therapeutic tools in the treatment of a variety of diseases.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty