Would you like to log in again?
Anti-Desmin Mouse Monoclonal Antibody [clone: D33]
Supplier: Diagnostic BioSystems
PDM006-RUO PDM006
76634-264EA 244.84 USD InStock
CA76634-264 76634-264
Anti-Desmin Mouse Monoclonal Antibody [clone: D33]
Antibodies
This antibody reacts with desmin protein of 53 kDa. Both striated as well as smooth muscle cells stain well with this antibody. The antibody shows a broad interspecific cross-reactivity, reacting with human.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty