Would you like to log in again?
Anti-CD31 (PECAM-1) Mouse Monoclonal Antibody [clone: JC/70A]
Supplier: Diagnostic BioSystems
PDM020-RUO PDM020
76634-286EA 286.64 USD InStock
CA76634-286 76634-286
Anti-CD31 (PECAM-1) Mouse Monoclonal Antibody [clone: JC/70A]
Antibodies
This antibody reacts with a 100 kDa glycoprotein expressed by endothelial cells and 130 kDa glycoprotein present in platelets. It stains endothelial cells in normal as well as malignant tissues.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty