Would you like to log in again?
Anti-Vimentin Mouse Monoclonal Antibody [clone: V9]
Supplier: Diagnostic BioSystems
PDM029-RUO PDM029
76634-290EA 252 USD InStock
76634-290 CA76634-290
Anti-Vimentin Mouse Monoclonal Antibody [clone: V9]
Antibodies
This antibody reacts with the 57 kDa intermediate filament protein present in the cells of mesenchymal origin. The antibody does not show any cross-reactivity with other intermediate filament proteins. However, it shows a broad interspecies cross-reactivity.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty