Would you like to log in again?
Anti-Alpha Fetoprotein Mouse Monoclonal Antibody [clone: C3]
Supplier: Diagnostic BioSystems
PDM057 PDM057-RUO
76634-314EA 254.39 USD InStock
76634-314 CA76634-314
Anti-Alpha Fetoprotein Mouse Monoclonal Antibody [clone: C3]
Antibodies
Alpha fetoprotein (AFP) is a single chain glycoprotein of 70 kDa that is normally expressed in the fetal liver, gastrointestinal tract and yolk sac. This antibody recognizes an epitope of α-fetoprotein present in human and several other species.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty