Would you like to log in again?
Anti-CD61 Platelet Glycoprotein IIIa Mouse Monoclonal Antibody [clone: Y2/51]
Supplier: Diagnostic BioSystems
PDM064 PDM064-RUO
76634-318EA 463.39 USD InStock
76634-318 CA76634-318
Anti-CD61 Platelet Glycoprotein IIIa Mouse Monoclonal Antibody [clone: Y2/51]
Antibodies
This antibody recognizes a glycoprotein of 110 kDa molecular weight present on human platelets and also reacts with megakaryocytes and megakaryoblasts. The antigen is also present on endothelium and osteoclasts. The glycoprotein IIIa molecule produced by endothelium cells is identical in sequence to platelet glycoprotein IIIa.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty