Would you like to log in again?
Anti-CD68 Macrophage Mouse Monoclonal Antibody [clone: PG-M1]
Supplier: Diagnostic BioSystems
PDM065-RUO PDM065
76634-320EA 440.69 USD InStock
76634-320 CA76634-320
Anti-CD68 Macrophage Mouse Monoclonal Antibody [clone: PG-M1]
Antibodies
This antibody reacts with a glycoprotein of 110 kDa, expressed as intracytoplasmic molecule. It stains macrophages in a wide variety of tissues. Myeloid precursor cells and peripheral granulocytes are negative. The PG-M1 differs from EMB11 (CD68) because of its non-reactivity with granulocytes and their precursor cells.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty