Would you like to log in again?
Anti-Cytokeratin 8/18 Mouse Monoclonal Antibody [clone: 5D3]
Supplier: Diagnostic BioSystems
PDM070 PDM070-RUO
76634-326EA 269.92 USD InStock
CA76634-326 76634-326
Anti-Cytokeratin 8/18 Mouse Monoclonal Antibody [clone: 5D3]
Antibodies
This antibody is specific to human cytokeratins 8 (52.5 kDa) and 18 (45 kDa). This antibody stains simple and glandular epithelium. This antibody can be useful for the identification of adenocarcinomas and most squamous cell carcinomas.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty