Would you like to log in again?
Anti-Cytokeratin AE1+AE3 Mouse Monoclonal Antibody [clone: AE1+AE3]
Supplier: Diagnostic BioSystems
PDM072-RUO PDM072
76634-328EA 228.12 USD InStock
76634-328 CA76634-328
Anti-Cytokeratin AE1+AE3 Mouse Monoclonal Antibody [clone: AE1+AE3]
Antibodies
This antibody recognizes cytokeratin polypeptides ranging from 40 to 58 kDa, corresponding to cytokeratins 5, 6, 8,17 and 19. It shows a broad pattern of reactivity with human epithelial tissues from simple glandular epithelia to stratified squamous epithelia, like epidermis, mammary gland ducts and tracheal epithelium.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty