Would you like to log in again?
Anti-Myosin Skeletal (Fast) Mouse Monoclonal Antibody [clone: MY32]
Supplier: Diagnostic BioSystems
PDM085 PDM085-RUO
76634-338EA 269.92 USD InStock
CA76634-338 76634-338
Anti-Myosin Skeletal (Fast) Mouse Monoclonal Antibody [clone: MY32]
Antibodies
This antibody recognizes heavy chain of human skeletal myosin. It does not stain cardiac or smooth muscle myosin. This antibody stains the fast twitch (type II) isomyosin molecule.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty