Would you like to log in again?
Anti-S-100 Mouse Monoclonal Antibody [clone: SH-B1]
Supplier: Diagnostic BioSystems
PDM088 PDM088-RUO
76634-340EA 451.44 USD InStock
CA76634-340 76634-340
Anti-S-100 Mouse Monoclonal Antibody [clone: SH-B1]
Antibodies
This antibody is specific against an epitope located on the β chain (i.e., in S-100a and S-100b) but not on the α chain of S-100 (i.e., in S-100a and S-100ao). This antibody can be used to localize S-100a and S-100b in various tissue sections.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty