Would you like to log in again?
Anti-CD8 T cell Mouse Monoclonal Antibody [clone: 144B]
Supplier: Diagnostic BioSystems
PDM094 PDM094-RUO
76634-342EA 309.32 USD InStock
76634-342 CA76634-342
Anti-CD8 T cell Mouse Monoclonal Antibody [clone: 144B]
Antibodies
This antibody reacts specifically with a 32 kDa protein similar to the CD8 molecule. This antibody stains cytotoxic/suppressor T cells and shows staining patterns similar to clustered CD8 monoclonal antibodies.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty