Would you like to log in again?
Anti-Actin sarcomeric Mouse Monoclonal Antibody [clone: 5C5]
Supplier: Diagnostic BioSystems
PDM099 PDM099-RUO
76634-348EA 299.77 USD InStock
CA76634-348 76634-348
Anti-Actin sarcomeric Mouse Monoclonal Antibody [clone: 5C5]
Antibodies
This antibody is specific to α-smooth muscle isoform of actin. It reacts with smooth muscle cells of vessels and different parenchymes. This antibody does not cross-react with β and γ-cytoplasmic, α-sarcomeric and α-myocardial actin isoforms.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty